1-(4-(Morpholinomethyl)phenyl)ethanone ≥97%

Lieferant: Apollo Scientific

OR510035-250MG
APOSOR510035-250MGEA 231 EUR
APOSOR510035-250MG
1-(4-(Morpholinomethyl)phenyl)ethanone ≥97%
1-(4-(Morpholinomethyl)phenyl)ethanone
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219.28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26679379
CAS-Nummer: 265107-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR