4-Bromo-1H-indole-7-carbonitrile ≥97%

Lieferant: Apollo Scientific

OR510036-250MG
APOSOR510036-250MGEA 144 EUR
APOSOR510036-250MG
4-Bromo-1H-indole-7-carbonitrile ≥97%
4-Bromo-1H-indole-7-carbonitrile
Formel: C₉H₅BrN₂
Molecular Weight: 221,053 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11845629
CAS-Nummer: 1167055-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR