3-Bromobenzo[b]thiophen-5-amine ≥98%
Lieferant: Apollo Scientific
OR510038-250MG
APOSOR510038-250MGEA
224
EUR
APOSOR510038-250MG
3-Bromobenzo[b]thiophen-5-amine ≥98%
3-Bromobenzo[b]thiophen-5-amine
Formel:
C₈H₆BrNS Molecular Weight: 228,109 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
98434-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...