5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-1,3-benzodioxole-4-carboxaldehyde

OR510039-1G
APOSOR510039-1GEA 72.5 EUR
APOSOR510039-1G
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde
Formel: C₈H₅BrO₃
Molecular Weight: 229,027 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02069012
CAS-Nummer: 72744-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR