Methyl 2-(3-bromopyridin-4-yl)acetate ≥97%

Lieferant: Apollo Scientific

OR510040-250MG
APOSOR510040-250MGEA 197 EUR
APOSOR510040-250MG
Methyl 2-(3-bromopyridin-4-yl)acetate ≥97%
Methyl 2-(3-bromopyridin-4-yl)acetate
Formel: C₈H₈BrNO₂
Molecular Weight: 230,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000071
CAS-Nummer: 162615-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR