4-Bromo-2-chloronicotinic acid > 97%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-chloro-nicotinic acid
OR510045-250MG
APOSOR510045-250MGEA
30.6
EUR
APOSOR510045-250MG
4-Bromo-2-chloronicotinic acid > 97%
4-Bromo-2-chloronicotinic acid
Formel:
C₆H₃BrClNO₂ Molecular Weight: 236.451 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD13189211 CAS-Nummer: 1060805-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...