(5-Bromo-2-methylphenyl)hydrazine hydrochloride ≥95%

Lieferant: Apollo Scientific

OR510046-250MG
APOSOR510046-250MGEA 122 EUR
APOSOR510046-250MG
(5-Bromo-2-methylphenyl)hydrazine hydrochloride ≥95%
(5-Bromo-2-methylphenyl)hydrazine hydrochloride
Formel: C₇H₉BrN₂·ClH
Molecular Weight: 237,525 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09834066
CAS-Nummer: 214915-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR