7-Bromo-4-methoxyquinazoline ≥98%

Lieferant: Apollo Scientific

OR510047-250MG
APOSOR510047-250MGEA 71.2 EUR
APOSOR510047-250MG
7-Bromo-4-methoxyquinazoline ≥98%
7-Bromo-4-methoxyquinazoline
Formel: C₉H₇BrN₂O
Molecular Weight: 239,069 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30834447
CAS-Nummer: 2140305-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR