4,6-Dibromopicolinonitrile ≥95%
Lieferant: Apollo Scientific
OR510050-250MG
APOSOR510050-250MGEA
0
EUR
APOSOR510050-250MG
4,6-Dibromopicolinonitrile ≥95%
4,6-Dibromopicolinonitrile
Formel:
C₆H₂Br₂N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD14582070 CAS-Nummer: 1206247-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...