3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine ≥97%
Lieferant: Apollo Scientific
OR510051-250MG
APOSOR510051-250MGEA
139
EUR
APOSOR510051-250MG
3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine ≥97%
3-Iodo-6-methoxy-1H-pyrrolo[3,2-c]pyridine
Formel:
C₈H₇IN₂O Molecular Weight: 274,058 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12963241 CAS-Nummer: 1190315-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...