5-Iodo-1H-indole-3-carboxylic acid ≥97%

Lieferant: Apollo Scientific

OR510052-250MG
APOSOR510052-250MGEA 140 EUR
APOSOR510052-250MG
5-Iodo-1H-indole-3-carboxylic acid ≥97%
5-Iodo-1H-indole-3-carboxylic acid
Formel: C₉H₆INO₂
Molecular Weight: 287,054 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15474905
CAS-Nummer: 1190847-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR