3-Chloro-2-iodo-5,6-dimethoxypyridine ≥98%
Lieferant: Apollo Scientific
OR510053-250MG
APOSOR510053-250MGEA
154
EUR
APOSOR510053-250MG
3-Chloro-2-iodo-5,6-dimethoxypyridine ≥98%
3-Chloro-2-iodo-5,6-dimethoxypyridine
Formel:
C₇H₇ClINO₂ Molecular Weight: 299,493 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22056183 CAS-Nummer: 910616-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...