5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde ≥95%

Lieferant: Apollo Scientific

OR510055-250MG
APOSOR510055-250MGEA 313 EUR
APOSOR510055-250MG
5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde ≥95%
5-Cyclopropyl-1-methyl-1H-1,2,4-triazole-3-carbaldehyde
Formel: C₇H₉N₃O
Molecular Weight: 151,166 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 877133-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR