3-Amino-6-chloropyridine-2(1H)-thione ≥97%

Lieferant: Apollo Scientific

OR510056-250MG
APOSOR510056-250MGEA 0 EUR
APOSOR510056-250MG
3-Amino-6-chloropyridine-2(1H)-thione ≥97%
3-Amino-6-chloropyridine-2(1H)-thione
Formel: C₅H₅ClN₂S
Lagertemperatur: Kühlschrank
MDL: MFCD22627872
CAS-Nummer: 27467-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR