4-(Dimethylamino)nicotinic acid ≥97%

Lieferant: Apollo Scientific

OR510058-250MG
APOSOR510058-250MGEA 127 EUR
APOSOR510058-250MG
4-(Dimethylamino)nicotinic acid ≥97%
4-(Dimethylamino)nicotinic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20654938
CAS-Nummer: 83039-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR