2-Chloro-6-methyl-4-nitropyridine > 98%

Lieferant: Apollo Scientific

OR510059-1G
APOSOR510059-1GEA 43.5 EUR
APOSOR510059-1G
2-Chloro-6-methyl-4-nitropyridine > 98%
2-Chloro-6-methyl-4-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.569 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD14582081
CAS-Nummer: 79055-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR