10-Bromo-7H-benzo[c]carbazole 99%
Lieferant: Apollo Scientific
OR51006-5G
APOSOR51006-5GEA
68.3
EUR
APOSOR51006-5G
10-Bromo-7H-benzo[c]carbazole 99%
10-Bromo-7H-benzo[c]carbazole
Formel:
C₁₆H₁₀BrN Molecular Weight: 296.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22054961 CAS-Nummer: 1698-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...