3-Bromo-1,2,4-thiadiazol-5-amine > 95%

Lieferant: Apollo Scientific

OR510060-250MG
APOSOR510060-250MGEA 237 EUR
APOSOR510060-250MG
3-Bromo-1,2,4-thiadiazol-5-amine > 95%
3-Bromo-1,2,4-thiadiazol-5-amine
Formel: C₂H₂BrN₃S
Molecular Weight: 180,026 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD13193591
CAS-Nummer: 1101173-93-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR