2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine > 95%

Lieferant: Apollo Scientific

OR510061-250MG
APOSOR510061-250MGEA 377 EUR
APOSOR510061-250MG
2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine > 95%
2-Chloro-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₇H₆ClN₃O
Molecular Weight: 183,595 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD14702347
CAS-Nummer: 96022-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR