5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine > 97%
Lieferant: Apollo Scientific
OR510062-250MG
APOSOR510062-250MGEA
302
EUR
APOSOR510062-250MG
5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine > 97%
5-Chloro-7-methoxypyrazolo[1,5-a]pyrimidine
Formel:
C₇H₆ClN₃O Molecular Weight: 183.595 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD20258250 CAS-Nummer: 1824286-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...