2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%

Lieferant: Apollo Scientific

OR510063-250MG
APOSOR510063-250MGEA 128 EUR
APOSOR510063-250MG
2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₈H₅ClN₂O₂
Molecular Weight: 196,59 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 522604-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR