3,5-Dichlorothiophene-2-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510064-250MG
APOSOR510064-250MGEA 108 EUR
APOSOR510064-250MG
3,5-Dichlorothiophene-2-carboxylic acid ≥98%
3,5-Dichlorothiophene-2-carboxylic acid
Formel: C₅H₂Cl₂O₂S
Molecular Weight: 197,039 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20638788
CAS-Nummer: 89166-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR