3-Bromo-2-methylbenzofuran ≥97%

Lieferant: Apollo Scientific

OR510065-250MG
APOSOR510065-250MGEA 173 EUR
APOSOR510065-250MG
3-Bromo-2-methylbenzofuran ≥97%
3-Bromo-2-methylbenzofuran
Formel: C₉H₇BrO
Molecular Weight: 211,055 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 58863-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR