2-(Bromomethyl)-5-nitropyridine > 97%

Lieferant: Apollo Scientific

OR510067-250MG
APOSOR510067-250MGEA 302 EUR
APOSOR510067-250MG
2-(Bromomethyl)-5-nitropyridine > 97%
2-(Bromomethyl)-5-nitropyridine
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09029365
CAS-Nummer: 887588-20-7
UN: 2810
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR