5-Bromo-2-(methylsulphinyl)pyrimidine ≥97%

Lieferant: Apollo Scientific

OR510068-250MG
APOSOR510068-250MGEA 43.5 EUR
APOSOR510068-250MG
5-Bromo-2-(methylsulphinyl)pyrimidine ≥97%
5-Bromo-2-(methylsulphinyl)pyrimidine
Formel: C₅H₅BrN₂OS
Molecular Weight: 221,075 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19689658
CAS-Nummer: 79685-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR