3-Amino-4-bromothiophene-2-carboxylic acid ≥98%
Lieferant: Apollo Scientific
OR510069-250MG
APOSOR510069-250MGEA
88.6
EUR
APOSOR510069-250MG
3-Amino-4-bromothiophene-2-carboxylic acid ≥98%
3-Amino-4-bromothiophene-2-carboxylic acid
Formel:
C₅H₄BrNO₂S Molecular Weight: 222,06 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
215927-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...