3-Bromonaphthalen-1-ol ≥97%

Lieferant: Apollo Scientific

OR510070-250MG
APOSOR510070-250MGEA 130 EUR
APOSOR510070-250MG
3-Bromonaphthalen-1-ol ≥97%
3-Bromonaphthalen-1-ol
Formel: C₁₀H₇BrO
Molecular Weight: 223,066 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17012415
CAS-Nummer: 90767-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR