5-Bromo-7-methylimidazo[5,1-f][1,2,4]triazin-4(1H)-one ≥98%
Lieferant: Apollo Scientific
OR510071-250MG
APOSOR510071-250MGEA
589
EUR
APOSOR510071-250MG
5-Bromo-7-methylimidazo[5,1-f][1,2,4]triazin-4(1H)-one ≥98%
5-Bromo-7-methylimidazo[5,1-f][1,2,4]triazin-4(1H)-one
Formel:
C₆H₅BrN₄O Molecular Weight: 229,034 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
1636161-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...