2-amino-5-bromo-1H-indole-3-carbonitrile ≥96%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-bromo-1H-indole-3-carbonitrile

OR510072-250MG
APOSOR510072-250MGEA 163 EUR
APOSOR510072-250MG
2-amino-5-bromo-1H-indole-3-carbonitrile ≥96%
2-amino-5-bromo-1H-indole-3-carbonitrile
Formel: C₉H₆BrN₃
Molecular Weight: 236,068 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20227060
CAS-Nummer: 1242140-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR