Ethyl 4-chloro-7-methyl-1H-indole-2-carboxylate > 95%

Lieferant: Apollo Scientific

OR510073-250MG
APOSOR510073-250MGEA 154 EUR
APOSOR510073-250MG
Ethyl 4-chloro-7-methyl-1H-indole-2-carboxylate > 95%
Ethyl 4-chloro-7-methyl-1H-indole-2-carboxylate
Formel: C₁₂H₁₂ClNO₂
Molecular Weight: 237,682 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD22556331
CAS-Nummer: 188248-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR