5-Iodo-1-methyluracil ≥96%

Lieferant: Apollo Scientific

OR510075-250MG
APOSOR510075-250MGEA 0 EUR
APOSOR510075-250MG
5-Iodo-1-methyluracil ≥96%
5-Iodo-1-methyluracil
Formel: C₅H₅IN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD22578662
CAS-Nummer: 45774-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR