Methyl 6-iodopyrazine-2-carboxylate ≥97%

Lieferant: Apollo Scientific

OR510076-250MG
APOSOR510076-250MGEA 161 EUR
APOSOR510076-250MG
Methyl 6-iodopyrazine-2-carboxylate ≥97%
Methyl 6-iodopyrazine-2-carboxylate
Formel: C₆H₅IN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD31618010
CAS-Nummer: 1603172-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR