6-Bromo-2-methyl-8-nitroquinoline ≥95%

Lieferant: Apollo Scientific

OR510077-1G
APOSOR510077-1GEA 168 EUR
APOSOR510077-1G
6-Bromo-2-methyl-8-nitroquinoline ≥95%
6-Bromo-2-methyl-8-nitroquinoline
Formel: C₁₀H₇BrN₂O₂
Molecular Weight: 267,079 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31630863
CAS-Nummer: 61854-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR