5-Bromo-2-morpholinonicotinonitrile ≥97%

Lieferant: Apollo Scientific

OR510078-1G
APOSOR510078-1GEA 34.8 EUR
APOSOR510078-1G
5-Bromo-2-morpholinonicotinonitrile ≥97%
5-Bromo-2-morpholinonicotinonitrile
Formel: C₁₀H₁₀BrN₃O
Molecular Weight: 268.11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21604970
CAS-Nummer: 1354223-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR