7-Bromo-5-chloro-8-methoxyquinoline ≥97%

Lieferant: Apollo Scientific

OR510079-250MG
APOSOR510079-250MGEA 87 EUR
APOSOR510079-250MG
7-Bromo-5-chloro-8-methoxyquinoline ≥97%
7-Bromo-5-chloro-8-methoxyquinoline
Formel: C₁₀H₇BrClNO
Lagertemperatur: Kühlschrank
MDL: MFCD22414761
CAS-Nummer: 457931-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR