2-Iodo-4-methoxy-1-(methylsulphonyl)benzene ≥97%

Lieferant: Apollo Scientific

OR510081-250MG
APOSOR510081-250MGEA 66.7 EUR
APOSOR510081-250MG
2-Iodo-4-methoxy-1-(methylsulphonyl)benzene ≥97%
2-Iodo-4-methoxy-1-(methylsulphonyl)benzene
Formel: C₈H₉IO₃S
Molecular Weight: 312,125 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30181850
CAS-Nummer: 1965309-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR