1H-Pyrazolo[4,3-c]pyridin-4-amine ≥95%

Lieferant: Apollo Scientific

OR510083-250MG
APOSOR510083-250MGEA 346 EUR
APOSOR510083-250MG
1H-Pyrazolo[4,3-c]pyridin-4-amine ≥95%
1H-Pyrazolo[4,3-c]pyridin-4-amine
Formel: C₆H₆N₄
Molecular Weight: 134,139 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1159829-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR