[1,2,4]Triazolo[4,3-a]pyrimidin-3-amine ≥98%
Lieferant: Apollo Scientific
OR510084-250MG
APOSOR510084-250MGEA
203
EUR
APOSOR510084-250MG
[1,2,4]Triazolo[4,3-a]pyrimidin-3-amine ≥98%
[1,2,4]Triazolo[4,3-a]pyrimidin-3-amine
Formel:
C₅H₅N₅ Molecular Weight: 135,127 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
1195625-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...