5-Amino-3-methoxypyrazine-2-carbonitrile ≥97%
Lieferant: Apollo Scientific
OR510086-250MG
APOSOR510086-250MGEA
154
EUR
APOSOR510086-250MG
5-Amino-3-methoxypyrazine-2-carbonitrile ≥97%
5-Amino-3-methoxypyrazine-2-carbonitrile
Formel:
C₆H₆N₄O Molecular Weight: 150,138 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD15144329 CAS-Nummer: 1137478-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...