Methyl 6-amino-2-methylnicotinate ≥97%
Lieferant: Apollo Scientific
OR510089-250MG
APOSOR510089-250MGEA
97.3
EUR
APOSOR510089-250MG
Methyl 6-amino-2-methylnicotinate ≥97%
Methyl 6-amino-2-methylnicotinate
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08062965 CAS-Nummer: 872355-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...