Methyl 6-amino-5-methylnicotinate ≥97%
Lieferant: Apollo Scientific
OR510090-250MG
APOSOR510090-250MGEA
224
EUR
APOSOR510090-250MG
Methyl 6-amino-5-methylnicotinate ≥97%
Methyl 6-amino-5-methylnicotinate
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12911637 CAS-Nummer: 1184913-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...