(3-Amino-4-nitro-phenyl)methanol ≥97%

Lieferant: Apollo Scientific

OR510091-250MG
APOSOR510091-250MGEA 53.8 EUR
APOSOR510091-250MG
(3-Amino-4-nitro-phenyl)methanol ≥97%
(3-Amino-4-nitro-phenyl)methanol
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18389932
CAS-Nummer: 37637-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR