6-Hydroxyimidazo[1,2-a]pyridine-2-carboxylic acid ≥95%

Lieferant: Apollo Scientific

OR510093-250MG
APOSOR510093-250MGEA 139 EUR
APOSOR510093-250MG
6-Hydroxyimidazo[1,2-a]pyridine-2-carboxylic acid ≥95%
6-Hydroxyimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₆N₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD31555414
CAS-Nummer: 1781183-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR