5-Chloro-N-methyl-3-nitropyridin-2-amine ≥97%

Lieferant: Apollo Scientific

OR510094-250MG
APOSOR510094-250MGEA 66.7 EUR
APOSOR510094-250MG
5-Chloro-N-methyl-3-nitropyridin-2-amine ≥97%
5-Chloro-N-methyl-3-nitropyridin-2-amine
Formel: C₆H₆ClN₃O₂
Molecular Weight: 187,584 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00222271
CAS-Nummer: 98142-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR