6-Methoxy-5-nitro-3-pyridinecarboxylic acid ≥97%

Lieferant: Apollo Scientific

OR510095-250MG
APOSOR510095-250MGEA 120 EUR
APOSOR510095-250MG
6-Methoxy-5-nitro-3-pyridinecarboxylic acid ≥97%
6-Methoxy-5-nitro-3-pyridinecarboxylic acid
Formel: C₇H₆N₂O₅
Molecular Weight: 198,133 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18257299
CAS-Nummer: 1211516-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR