Methyl 2-cyano-5-nitrobenzoate ≥97%

Lieferant: Apollo Scientific

OR510096-250MG
APOSOR510096-250MGEA 94.4 EUR
APOSOR510096-250MG
Methyl 2-cyano-5-nitrobenzoate ≥97%
Methyl 2-cyano-5-nitrobenzoate
Formel: C₉H₆N₂O₄
Molecular Weight: 206,155 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11975694
CAS-Nummer: 1260777-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR