Methyl 3-amino-4-methyl-5-nitrobenzoate ≥98%

Lieferant: Apollo Scientific

OR510098-250MG
APOSOR510098-250MGEA 85.7 EUR
APOSOR510098-250MG
Methyl 3-amino-4-methyl-5-nitrobenzoate ≥98%
Methyl 3-amino-4-methyl-5-nitrobenzoate
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,187 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03081801
CAS-Nummer: 72922-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR