Ethyl 3-chloro-6-cyanopicolinate ≥95%

Lieferant: Apollo Scientific

OR510099-1G
APOSOR510099-1GEA 62.5 EUR
APOSOR510099-1G
Ethyl 3-chloro-6-cyanopicolinate ≥95%
Ethyl 3-chloro-6-cyanopicolinate
Formel: C₉H₇ClN₂O₂
Molecular Weight: 210,617 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28749724
CAS-Nummer: 1436504-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR