3-Amino-6-bromopicolinic acid ≥98%
Lieferant: Apollo Scientific
OR510100-250MG
APOSOR510100-250MGEA
102
EUR
APOSOR510100-250MG
3-Amino-6-bromopicolinic acid ≥98%
3-Amino-6-bromopicolinic acid
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217.02 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD14584498 CAS-Nummer: 1052708-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...