2-Amino-5-bromo-6-chloropyrimidin-4(1H)-one ≥95%

Lieferant: Apollo Scientific

OR510101-250MG
APOSOR510101-250MGEA 145 EUR
APOSOR510101-250MG
2-Amino-5-bromo-6-chloropyrimidin-4(1H)-one ≥95%
2-Amino-5-bromo-6-chloropyrimidin-4(1H)-one
Formel: C₄H₃BrClN₃O
Molecular Weight: 224,443 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 55740-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR